Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC VU0424465 50mg | 1428630-85-6 | 50 MG
SDP

Catalog No. 5000226895
Encompass_Preferred

VU0424465 is a research small molecule characterized as a potent, partial positive allosteric modulator (PAM)-agonist of the metabotropic glutamate receptor 5 (mGlu5). It is supplied for laboratory research use only and is not intended for human consumption.

  • High reported purity of 99.84%.
  • Molecular weight 326.36 g/mol.
  • Chemical formula C19H19FN2O2.
  • Available in small pack sizes such as 50 mg for assay development.
  • Suitable for pharmacology and medicinal chemistry research.

Catalog No. 50-002-26895 Supplier Medchemexpress LLC Supplier No. HY11497850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VU0424465 is a research small molecule characterized as a potent, partial positive allosteric modulator (PAM)-agonist of the metabotropic glutamate receptor 5 (mGlu5). It is supplied for laboratory research use only and is not intended for human consumption.

  • High reported purity of 99.84%.
  • Molecular weight 326.36 g/mol.
  • Chemical formula C19H19FN2O2.
  • Available in small pack sizes such as 50 mg for assay development.
  • Suitable for pharmacology and medicinal chemistry research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.