Learn More
Medchemexpress LLC VU0453379 hydrochloride | 00-00-0 | MFCD28987471 | 99.0% | 471.03 g·mol⁻¹ | C26H35ClN4O2 | 5 MG
SDP
Catalog No. 5000264574
Encompass_Preferred
VU0453379 hydrochloride is a selective, CNS-penetrant positive allosteric modulator of the glucagon-like peptide-1 receptor (GLP-1R) used as a research reagent to study GLP-1R pharmacology. It potentiates GLP-1R signaling with an EC50 of approximately 1.3 μM and is supplied as a high-purity hydrochloride salt for in vitro and in vivo studies.
  • Selective GLP-1R positive allosteric modulator with EC50 ~1.3 μM.
  • CNS-penetrant compound suitable for central nervous system studies.
  • High purity (listed as 99.0%) for reproducible research results.
  • Provided as hydrochloride salt with molecular weight 471.03 g·mol⁻¹.
  • Available in small research pack sizes and DMSO solutions for assay use.
  • Store at 4°C, protected from moisture.
Catalog No. 50-002-64574 Supplier Medchemexpress LLC Supplier No. HY116819A5MG
VU0453379 hydrochloride is a selective, CNS-penetrant positive allosteric modulator of the glucagon-like peptide-1 receptor (GLP-1R) used as a research reagent to study GLP-1R pharmacology. It potentiates GLP-1R signaling with an EC50 of approximately 1.3 μM and is supplied as a high-purity hydrochloride salt for in vitro and in vivo studies.
  • Selective GLP-1R positive allosteric modulator with EC50 ~1.3 μM.
  • CNS-penetrant compound suitable for central nervous system studies.
  • High purity (listed as 99.0%) for reproducible research results.
  • Provided as hydrochloride salt with molecular weight 471.03 g·mol⁻¹.
  • Available in small research pack sizes and DMSO solutions for assay use.
  • Store at 4°C, protected from moisture.