Learn More
Medchemexpress LLC VU0453379 hydrochloride | 1638646-27-1 | 99.0% | 471.03 | 100 MG
SDP
Catalog No. 5000437152
Encompass_Preferred
VU0453379 hydrochloride is a highly selective and central nervous system (CNS) penetrant positive allosteric modulator (PAM) of glucagon-like peptide-1R (GLP-1R) with an EC50 of 1.3 μM. It is for research use only and not sold to patients. The product is described as a highly selective and CNS penetrant positive allosteric modulator.
  • Highly selective
  • CNS penetrant positive allosteric modulator
Catalog No. 50-004-37152 Supplier Medchemexpress LLC Supplier No. HY116819A100MG
VU0453379 hydrochloride is a highly selective and central nervous system (CNS) penetrant positive allosteric modulator (PAM) of glucagon-like peptide-1R (GLP-1R) with an EC50 of 1.3 μM. It is for research use only and not sold to patients. The product is described as a highly selective and CNS penetrant positive allosteric modulator.
  • Highly selective
  • CNS penetrant positive allosteric modulator