Learn More
Medchemexpress LLC VU0453379 hydrochloride | 98.0% | 471.03 | 25 MG
SDP
Catalog No. 5000438300
Encompass_Preferred
VU0453379 hydrochloride is a highly selective and central nervous system (CNS) penetrant positive allosteric modulator (PAM) of glucagon-like peptide-1R (GLP-1R) with an EC50 of 1.3 μM.
  • Highly selective
  • CNS penetrant
  • Positive allosteric modulator (PAM) of GLP-1R
Catalog No. 50-004-38300 Supplier Medchemexpress LLC Supplier No. 25MGHY116819A
VU0453379 hydrochloride is a highly selective and central nervous system (CNS) penetrant positive allosteric modulator (PAM) of glucagon-like peptide-1R (GLP-1R) with an EC50 of 1.3 μM.
  • Highly selective
  • CNS penetrant
  • Positive allosteric modulator (PAM) of GLP-1R