Learn More
Medchemexpress LLC VU0453379 hydrochloride | 99.0% | 471.03 | 1 ML
SDP
Catalog No. 5000435578
Encompass_Preferred
VU0453379 hydrochloride is a highly selective and central nervous system (CNS) penetrant positive allosteric modulator (PAM) of glucagon-like peptide-1R (GLP-1R). It exhibits an EC50 of 1.3 μM.
  • Highly selective positive allosteric modulator.
  • Central nervous system (CNS) penetrant.
  • Modulates glucagon-like peptide-1R (GLP-1R).
  • Demonstrates an EC50 of 1.3 μM.
Catalog No. 50-004-35578 Supplier Medchemexpress LLC Supplier No. HY116819A10MM/1ML
VU0453379 hydrochloride is a highly selective and central nervous system (CNS) penetrant positive allosteric modulator (PAM) of glucagon-like peptide-1R (GLP-1R). It exhibits an EC50 of 1.3 μM.
  • Highly selective positive allosteric modulator.
  • Central nervous system (CNS) penetrant.
  • Modulates glucagon-like peptide-1R (GLP-1R).
  • Demonstrates an EC50 of 1.3 μM.