Learn More
Medchemexpress LLC VU0453379 (hydrochloride) | 99.0% | 471.03 | 10 MG
SDP
Catalog No. 5000435577
Encompass_Preferred
VU0453379 hydrochloride is a highly selective and central nervous system (CNS) penetrant positive allosteric modulator (PAM) of glucagon-like peptide-1R (GLP-1R) with an EC50 of 1.3 μM.
  • Highly selective
  • Central nervous system (CNS) penetrant
  • Positive allosteric modulator (PAM) of glucagon-like peptide-1R (GLP-1R)
Catalog No. 50-004-35577 Supplier Medchemexpress LLC Supplier No. HY116819A10MG
VU0453379 hydrochloride is a highly selective and central nervous system (CNS) penetrant positive allosteric modulator (PAM) of glucagon-like peptide-1R (GLP-1R) with an EC50 of 1.3 μM.
  • Highly selective
  • Central nervous system (CNS) penetrant
  • Positive allosteric modulator (PAM) of glucagon-like peptide-1R (GLP-1R)