Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC VU0453379 hydrochloride | 99.0% | 471.03 | 50 MG
SDP

Catalog No. 5000435575
Encompass_Preferred

VU0453379 hydrochloride is a highly selective and central nervous system (CNS) penetrant positive allosteric modulator (PAM) of glucagon-like peptide-1R (GLP-1R) with an EC50 of 1.3 μM. It is for research use only.

  • Purity: 99.0%
  • Molecular weight: 471.03
  • Formula: C26H35ClN4O2
  • Appearance: Solid
  • Color: White to off-white

Catalog No. 50-004-35575 Supplier Medchemexpress LLC Supplier No. HY116819A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VU0453379 hydrochloride is a highly selective and central nervous system (CNS) penetrant positive allosteric modulator (PAM) of glucagon-like peptide-1R (GLP-1R) with an EC50 of 1.3 μM. It is for research use only.

  • Purity: 99.0%
  • Molecular weight: 471.03
  • Formula: C26H35ClN4O2
  • Appearance: Solid
  • Color: White to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.