Learn More
Medchemexpress LLC VU0463271 | 1391737-01-1 | 99.8% | 382.50 | 1 ML
SDP
Catalog No. 5000344321
Encompass_Preferred
VU0463271 is a selective KCC2 antagonist, with an IC50 of 61 nM. It offers a potent and selective antagonist activity, making it suitable for various research applications.
  • Selective KCC2 antagonist
  • IC50 of 61 nM (KCC2)
  • Greater than 100-fold selectivity versus the closely related Na-K-2Cl cotransporter 1 (NKCC1)
  • No activity in a larger panel of GPCRs, ion channels, and transporters
  • Rapidly cleared in vitro
Catalog No. 50-003-44321 Supplier Medchemexpress LLC Supplier No. HY110110100MG
VU0463271 is a selective KCC2 antagonist, with an IC50 of 61 nM. It offers a potent and selective antagonist activity, making it suitable for various research applications.
  • Selective KCC2 antagonist
  • IC50 of 61 nM (KCC2)
  • Greater than 100-fold selectivity versus the closely related Na-K-2Cl cotransporter 1 (NKCC1)
  • No activity in a larger panel of GPCRs, ion channels, and transporters
  • Rapidly cleared in vitro