Learn More
Medchemexpress LLC HY-112209 5mg Medchemexpress, VU0467154 CAS:1451993-15-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1122095MG
Encompass_Preferred
Medchemexpress, HY-112209 5mg VU0467154 CAS:1451993-15-9 VU0467154 is a positive allosteric modulator of the M4 muscarinic acetylcholine receptor (mAChR), potentiating the response to ACh with pEC50s of 7.75, 6.2 and 6 for rat, human and cynomolgus monkey M4 receptor, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9060