Learn More
Medchemexpress LLC Vu0467485 | 1451994-10-7 | 99.3% | 25 MG
SDP
Catalog No. 5000234379
Encompass_Preferred
VU0467485 (AZ13713945) is a potent, selective, orally bioavailable muscarinic acetylcholine receptor 4 (M4) positive allosteric modulator supplied for research use.
  • Potent and selective M4 positive allosteric modulator
  • Orally bioavailable in preclinical studies
  • High purity suitable for analytical and screening applications
  • Available in small milligram-scale packages for research
  • Characterized molecular weight and formula for reproducibility
Catalog No. 50-002-34379 Supplier Medchemexpress LLC Supplier No. HY12018425MG
VU0467485 (AZ13713945) is a potent, selective, orally bioavailable muscarinic acetylcholine receptor 4 (M4) positive allosteric modulator supplied for research use.
  • Potent and selective M4 positive allosteric modulator
  • Orally bioavailable in preclinical studies
  • High purity suitable for analytical and screening applications
  • Available in small milligram-scale packages for research
  • Characterized molecular weight and formula for reproducibility