Learn More
Medchemexpress LLC Vu0467485 (AZ13713945) | 1451994-10-7 | 99.3% | 10 MG
SDP
Catalog No. 5000234428
Encompass_Preferred
VU0467485 (AZ13713945) is a potent, selective, orally bioavailable positive allosteric modulator (PAM) of the muscarinic acetylcholine receptor M4 supplied as a solid research reagent for preclinical studies.
  • Potent and selective M4 receptor positive allosteric modulator.
  • Orally bioavailable in preclinical models.
  • High purity solid suitable for research applications.
  • Provided in small milligram pack sizes for preclinical dosing.
  • Molecular formula C17H17FN4O2S; molecular weight 360.41 g/mol.
Catalog No. 50-002-34428 Supplier Medchemexpress LLC Supplier No. HY12018410MG
VU0467485 (AZ13713945) is a potent, selective, orally bioavailable positive allosteric modulator (PAM) of the muscarinic acetylcholine receptor M4 supplied as a solid research reagent for preclinical studies.
  • Potent and selective M4 receptor positive allosteric modulator.
  • Orally bioavailable in preclinical models.
  • High purity solid suitable for research applications.
  • Provided in small milligram pack sizes for preclinical dosing.
  • Molecular formula C17H17FN4O2S; molecular weight 360.41 g/mol.