Learn More
Cayman Chemical VU0469650 (hydrochloride), Purity: ≥98%, 10 mg
SDP
Catalog No. 502949898
Encompass_Preferred
A negative allosteric mGluR1 modulator; inhibits glutamate-induced calcium flux in cells expressing human mGluR1 (IC50 = 99 nM); >100-fold selective for mGluR1 over mGluR2-8 at 10 µM
Catalog No. 50-294-9898 Supplier Cayman Chemical Supplier No. 3293310 MG
A negative allosteric mGluR1 modulator; inhibits glutamate-induced calcium flux in cells expressing human mGluR1 (IC50 = 99 nM); >100-fold selective for mGluR1 over mGluR2-8 at 10 µM