Learn More
TARGETMOL CHEMICALS INC VU0483605 5MG
SDP
Catalog No. 502581008
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. VU0483605 is an effective and selective positive allosteric modulator of mGluR1 with EC50s of 390 and 356 nM for human and rat respectively. purity: 97%
Catalog No. 50-258-1008 Supplier TARGETMOL CHEMICALS INC Supplier No. T217475MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. VU0483605 is an effective and selective positive allosteric modulator of mGluR1 with EC50s of 390 and 356 nM for human and rat respectively. purity: 97%