Learn More
Medchemexpress LLC HY-112705 1mg Medchemexpress, VU0529331 CAS:1286725-49-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1127051MG
Encompass_Preferred
Medchemexpress, HY-112705 1mg VU0529331 CAS:1286725-49-2 VU0529331 is a modestly selective non-GIRK1-containing G protein-gated, inwardly-rectifying, potassium channel (non-GIRK1/X) activator, with EC50s of 5.1 µM and 5.2 µM for GIRK2 and GIRK1/2 in HEK293 cells, respectively, also effective on GIRK4 homomeric channel. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9148