Learn More
Medchemexpress LLC HY-112705 5mg Medchemexpress, VU0529331 CAS:1286725-49-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1127055MG
Encompass_Preferred
Medchemexpress, HY-112705 5mg VU0529331 CAS:1286725-49-2 VU0529331 is a modestly selective non-GIRK1-containing G protein-gated, inwardly-rectifying, potassium channel ( non-GIRK1/X ) activator, with EC 50 s of 5.1 µM and 5.2 µM for GIRK2 and GIRK1/2 in HEK293 cells, respectively, also effective on GIRK4 homomeric channel [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2431