Learn More
Medchemexpress LLC HY-108710 5mg Medchemexpress, VU0650786 CAS:1809085-30-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1087105MG
Encompass_Preferred
Medchemexpress, HY-108710 5mg VU0650786 CAS:1809085-30-0 VU0650786 is a potent and selective CNS penetrant negative allosteric modulator of metabotropic glutamate receptor subtype 3 (mGlu3 NAM) , with an IC 50 of 392 nM. VU0650786 has antidepressant and anxiolytic activity in rodents [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2333