Learn More
Medchemexpress LLC HY-112859 10mg Medchemexpress, VU0661013 CAS:2131184-57-9 Purity:>98%
SDP
Catalog No. 501969191
Encompass_Preferred
Medchemexpress, HY-112859 10mg VU0661013 CAS:2131184-57-9 VU661013 is a potent and selective MCL-1 inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9191 Supplier Medchemexpress LLC Supplier No. HY11285910MG
Medchemexpress, HY-112859 10mg VU0661013 CAS:2131184-57-9 VU661013 is a potent and selective MCL-1 inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.