Learn More
Cayman Chemical VU590hydrochlorIde 10mg
SDP
Catalog No. 502973290
Encompass_Preferred
A small molecule inhibitor of KCNJ1 (IC50 = 294 nM) which also causes 70% inhibition of KCNJ13 at 10 µM; has no effect on the related channels KCNJ2 (Kir2.1) and KCNJ10 (Kir4.1) at a concentration of 10 µM
Catalog No. 50-297-3290 Supplier Cayman Chemical Supplier No. 1517710 MG
A small molecule inhibitor of KCNJ1 (IC50 = 294 nM) which also causes 70% inhibition of KCNJ13 at 10 µM; has no effect on the related channels KCNJ2 (Kir2.1) and KCNJ10 (Kir4.1) at a concentration of 10 µM