Learn More
Medchemexpress LLC HY-108585A 5mg Medchemexpress, VU591 CAS:1222810-74-3 Purity:>98%
SDP
Catalog No. 501932321
Encompass_Preferred
Medchemexpress, HY-108585A 5mg VU591 CAS:1222810-74-3 VU591 is a potent, selective renal outer medullary potassium channel (ROMK or Kir1.1) inhibitor, with an IC 50 of 0.24 μM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2321 Supplier Medchemexpress LLC Supplier No. HY108585A5MG
Medchemexpress, HY-108585A 5mg VU591 CAS:1222810-74-3 VU591 is a potent, selective renal outer medullary potassium channel (ROMK or Kir1.1) inhibitor, with an IC 50 of 0.24 μM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.