Learn More
Medchemexpress LLC HY-108585 5mg Medchemexpress, VU591 hydrochloride CAS:1315380-70-1 Purity:>98%
SDP
Catalog No. 501968580
Encompass_Preferred
Medchemexpress, HY-108585 5mg VU591 HCl CAS:1315380-70-1 VU591 hydrochloride is a potent, selective renal outer medullary potassium channel (ROMK or Kir1.1) inhibitor, with an IC50 of 0.24 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8580 Supplier Medchemexpress LLC Supplier No. HY1085855MG
Medchemexpress, HY-108585 5mg VU591 HCl CAS:1315380-70-1 VU591 hydrochloride is a potent, selective renal outer medullary potassium channel (ROMK or Kir1.1) inhibitor, with an IC50 of 0.24 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.