Learn More
Medchemexpress LLC HY-107982 5mg Medchemexpress, VU6005649 CAS:2137047-43-7 Purity:>98%
SDP
Catalog No. 501968488
Encompass_Preferred
Medchemexpress, HY-107982 5mg VU6005649 CAS:2137047-43-7 VU6005649 is a CNS penetrant mGlu7/8 receptor agonist with EC50s of 0.65 μM and 2.6 μM for mGlu7 receptor and mGlu8 receptor, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8488 Supplier Medchemexpress LLC Supplier No. HY1079825MG
Medchemexpress, HY-107982 5mg VU6005649 CAS:2137047-43-7 VU6005649 is a CNS penetrant mGlu7/8 receptor agonist with EC50s of 0.65 μM and 2.6 μM for mGlu7 receptor and mGlu8 receptor, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.