Learn More
TARGETMOL CHEMICALS INC VU6012962 10MG
SDP
Catalog No. 502577399
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. VU6012962 is an orally bioavailable negative allosteric modulator of CNS-penetrant metabotropic glutamate receptor 7(mGlu7; IC50 347 nM). purity: 98%
Catalog No. 50-257-7399 Supplier TARGETMOL CHEMICALS INC Supplier No. T1332310MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. VU6012962 is an orally bioavailable negative allosteric modulator of CNS-penetrant metabotropic glutamate receptor 7(mGlu7; IC50 347 nM). purity: 98%