Learn More
Medchemexpress LLC HY-114403 10mg Medchemexpress, VU6012962 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11440310MG
Encompass_Preferred
Medchemexpress, HY-114403 10mg VU6012962 CAS: VU6012962 is an orally bioavailable and CNS-penetrant metabotropic glutamate receptor 7 negative allosteric modulator (mGlu7 NAM) with an IC50 of 347 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9346