Learn More
Medchemexpress LLC HY-135401 5mg Medchemexpress, VU6015929 CAS: Purity:>98%
SDP
Catalog No. 501969875
Encompass_Preferred
Medchemexpress, HY-135401 5mg VU6015929 CAS: VU6015929 is a potent, selective and orally active dual discoidin domain receptor 1/2 (DDR1/2) inhibitor with IC50s of 4.67 nM and 7.39 nM, respectively. VU6015929 potently blocks collagen-induced DDR1 activation and collagen-IV production. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9875 Supplier Medchemexpress LLC Supplier No. HY1354015MG
Medchemexpress, HY-135401 5mg VU6015929 CAS: VU6015929 is a potent, selective and orally active dual discoidin domain receptor 1/2 (DDR1/2) inhibitor with IC50s of 4.67 nM and 7.39 nM, respectively. VU6015929 potently blocks collagen-induced DDR1 activation and collagen-IV production. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.