Learn More
Medchemexpress LLC Vu6036720 hydrochloride | 3026597-12-3 | 99.7% | 473.39 | C20H23Cl2FN4O2S | 10 MG
SDP
Catalog No. 5000287202
Encompass_Preferred
VU6036720 hydrochloride is the hydrochloride salt of a potent, selective in vitro inhibitor of heteromeric Kir4.1/5.1 inward-rectifier potassium channels, used for research into neural and renal channel function. The compound is supplied as a high-purity solid suitable for biochemical and electrophysiological assays.
  • Potent Kir4.1/5.1 inhibition with IC50 0.24 μM.
  • High purity suitable for research applications.
  • Available in small solid quantities for assay workflows.
  • Stable solid with recommended sealed storage at 4°C.
  • Compatible with DMSO stock preparations for in vitro testing.
Catalog No. 50-002-87202 Supplier Medchemexpress LLC Supplier No. HY148304A10MG
VU6036720 hydrochloride is the hydrochloride salt of a potent, selective in vitro inhibitor of heteromeric Kir4.1/5.1 inward-rectifier potassium channels, used for research into neural and renal channel function. The compound is supplied as a high-purity solid suitable for biochemical and electrophysiological assays.
  • Potent Kir4.1/5.1 inhibition with IC50 0.24 μM.
  • High purity suitable for research applications.
  • Available in small solid quantities for assay workflows.
  • Stable solid with recommended sealed storage at 4°C.
  • Compatible with DMSO stock preparations for in vitro testing.