Learn More
Medchemexpress LLC VU6036720 (hydrochloride) | 99.65% | 473.39 | 25 MG
SDP
Catalog No. 5000362735
Encompass_Preferred
VU6036720 hydrochloride is a potent and specific in vitro inhibitor of Kir4.1/5.1, effectively inhibiting these channels with an IC50 value of 0.24 μM. This compound is utilized in research focusing on brain and kidney functions.
  • Potent and specific in vitro inhibitor of Kir4.1/5.1 channels
  • Inhibits Kir4.1/5.1 channels with an IC50 value of 0.24 μM
  • Useful for research related to the brain and kidney
  • Storage conditions: 4°C, sealed storage, away from moisture and light. In solvent: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture and light).
Catalog No. 50-003-62735 Supplier Medchemexpress LLC Supplier No. 25MGHY148304A
VU6036720 hydrochloride is a potent and specific in vitro inhibitor of Kir4.1/5.1, effectively inhibiting these channels with an IC50 value of 0.24 μM. This compound is utilized in research focusing on brain and kidney functions.
  • Potent and specific in vitro inhibitor of Kir4.1/5.1 channels
  • Inhibits Kir4.1/5.1 channels with an IC50 value of 0.24 μM
  • Useful for research related to the brain and kidney
  • Storage conditions: 4°C, sealed storage, away from moisture and light. In solvent: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture and light).