Learn More
Medchemexpress LLC VU6036720 hydrochloride | 99.65% | 473.39 | 50 MG
SDP
Catalog No. 5000362317
Encompass_Preferred
VU6036720 hydrochloride is a potent and specific inhibitor of Kir4.1/5.1 channels. It is suitable for various research applications.
  • Inhibits Kir4.1/5.1 channels with an IC50 value of 0.24 μM.
  • Can be used for the research of brain and kidney.
Catalog No. 50-003-62317 Supplier Medchemexpress LLC Supplier No. HY148304A50MG
VU6036720 hydrochloride is a potent and specific inhibitor of Kir4.1/5.1 channels. It is suitable for various research applications.
  • Inhibits Kir4.1/5.1 channels with an IC50 value of 0.24 μM.
  • Can be used for the research of brain and kidney.