Learn More
Medchemexpress LLC VU6036720 hydrochloride | 99.7% | 473.39 | 5 MG
SDP
Catalog No. 5000362170
Encompass_Preferred
VU6036720 hydrochloride is a potent and specific in vitro inhibitor of Kir4.1/5.1. It can inhibit Kir4.1/5.1 channels with an IC50 value of 0.24 μM and can be used for the research of brain and kidney.
  • Potent and specific in vitro inhibitor of Kir4.1/5.1.
  • Inhibits Kir4.1/5.1 channels with an IC50 value of 0.24 μM.
  • Useful for research pertaining to the brain.
  • Useful for research pertaining to the kidney.
Catalog No. 50-003-62170 Supplier Medchemexpress LLC Supplier No. HY148304A5MG
VU6036720 hydrochloride is a potent and specific in vitro inhibitor of Kir4.1/5.1. It can inhibit Kir4.1/5.1 channels with an IC50 value of 0.24 μM and can be used for the research of brain and kidney.
  • Potent and specific in vitro inhibitor of Kir4.1/5.1.
  • Inhibits Kir4.1/5.1 channels with an IC50 value of 0.24 μM.
  • Useful for research pertaining to the brain.
  • Useful for research pertaining to the kidney.