Learn More
Accuflow Systems Saint-Gobain VueLife Closure Clamp, White, 22 cm, 1.2 cm
Catalog No. NC3454849
Encompass_Preferred
The VueLife closure clamp is a removable closure clamp designed to securely seal C-series bioprocess FEP bags used in cell culture and bioprocessing operations. It provides a simple mechanical closure for bag ports and tubing and can be repositioned to accommodate expanding cultures.
  • Closure clamp for FEP bioprocess bags
  • Length 22 cm
  • Width 1.2 cm
  • White color
  • Removable and repositionable
  • Suitable for aseptic bag handling in bioprocess workflows
Catalog No. NC3454849 Supplier Accuflow Systems Supplier No. 1C0022
The VueLife closure clamp is a removable closure clamp designed to securely seal C-series bioprocess FEP bags used in cell culture and bioprocessing operations. It provides a simple mechanical closure for bag ports and tubing and can be repositioned to accommodate expanding cultures.
  • Closure clamp for FEP bioprocess bags
  • Length 22 cm
  • Width 1.2 cm
  • White color
  • Removable and repositionable
  • Suitable for aseptic bag handling in bioprocess workflows