Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VUF 11207 fumarate, Tocris Bioscience™
Description
Potent ACKR3 (CXCR7) chemokine receptor agonist (EC50 = 1.6 nM). Induces recruitment of β-arrestin to CXCR7 in HEK293-CXCR7 cells.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 586.65 |
| Chemical Name or Material | VUF 11207 fumarate |
Safety and Handling
| Recommended Storage | Store at -20°C |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?