Learn More
Medchemexpress LLC VUF-5574 | 280570-45-8 | MFCD06691423 | 99.0% | 371.39 | C21H17N5O2 | 25 MG
SDP
Catalog No. 5000247279
Encompass_Preferred
VUF-5574 is a selective antagonist of the adenosine A3 receptor (A3R) with a reported Ki of 4.03 nM for the recombinant human receptor. It is provided as a research-grade small molecule (formula C21H17N5O2; MW 371.39) commonly used to study A3 receptor pharmacology and signaling.
  • Selective A3 receptor antagonist (Ki = 4.03 nM for recombinant human A3R).
  • High chemical purity (99.0%), suitable for in vitro research.
  • Characterized small molecule: formula C21H17N5O2, MW 371.39.
  • Available in multiple pack sizes for assay development and screening.
  • Intended for research use only, not for human therapeutic applications.
Catalog No. 50-002-47279 Supplier Medchemexpress LLC Supplier No. HY10318925MG
VUF-5574 is a selective antagonist of the adenosine A3 receptor (A3R) with a reported Ki of 4.03 nM for the recombinant human receptor. It is provided as a research-grade small molecule (formula C21H17N5O2; MW 371.39) commonly used to study A3 receptor pharmacology and signaling.
  • Selective A3 receptor antagonist (Ki = 4.03 nM for recombinant human A3R).
  • High chemical purity (99.0%), suitable for in vitro research.
  • Characterized small molecule: formula C21H17N5O2, MW 371.39.
  • Available in multiple pack sizes for assay development and screening.
  • Intended for research use only, not for human therapeutic applications.