Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC VUF-5574 | 280570-45-8 | MFCD06691423 | 99.0% | 371.39 g·mol⁻¹ | C21H17N5O2 | 50 MG
SDP

Catalog No. 5000246706
Encompass_Preferred

VUF-5574 is a small-molecule research compound that acts as a selective antagonist of the adenosine A3 (A3R) receptor, with a reported Ki of 4.03 nM for the recombinant human receptor. It is supplied as a high-purity solid intended for in vitro pharmacology and receptor profiling; for research use only.

  • Selective adenosine A3 receptor antagonist (Ki = 4.03 nM).
  • Molecular formula C21H17N5O2; molecular weight 371.39 g·mol⁻¹.
  • Purity 99.0% as supplied.
  • Available in small-scale packages suitable for laboratory research (e.g., 50 MG).
  • Provides supporting documentation such as COA, SDS, and spectral data.

Catalog No. 50-002-46706 Supplier Medchemexpress LLC Supplier No. HY10318950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VUF-5574 is a small-molecule research compound that acts as a selective antagonist of the adenosine A3 (A3R) receptor, with a reported Ki of 4.03 nM for the recombinant human receptor. It is supplied as a high-purity solid intended for in vitro pharmacology and receptor profiling; for research use only.

  • Selective adenosine A3 receptor antagonist (Ki = 4.03 nM).
  • Molecular formula C21H17N5O2; molecular weight 371.39 g·mol⁻¹.
  • Purity 99.0% as supplied.
  • Available in small-scale packages suitable for laboratory research (e.g., 50 MG).
  • Provides supporting documentation such as COA, SDS, and spectral data.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.