Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Vuf 8430 dihydrobromide | 100130-32-3 | 98.0% | 323.05 g/mol | C4H13Br2N5S | 10 MG
SDP

Catalog No. 5000240382
Encompass_Preferred

VUF 8430 dihydrobromide is a potent, selective histamine H4 receptor agonist supplied as a solid research reagent. It exhibits a Ki of 31.6 nM and an EC50 of 50 nM at the H4 receptor, and is provided in small milligram quantities for pharmacological and biochemical studies. For research use only.

  • Purity 98.0%
  • Molecular weight 323.05 g/mol
  • CAS 100130-32-3
  • Appearance solid
  • Potency Ki = 31.6 nM; EC50 = 50 nM (histamine H4 receptor)
  • Available in small milligram pack sizes, including 10 mg
  • Intended for research use only

Catalog No. 50-002-40382 Supplier Medchemexpress LLC Supplier No. HY10755510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VUF 8430 dihydrobromide is a potent, selective histamine H4 receptor agonist supplied as a solid research reagent. It exhibits a Ki of 31.6 nM and an EC50 of 50 nM at the H4 receptor, and is provided in small milligram quantities for pharmacological and biochemical studies. For research use only.

  • Purity 98.0%
  • Molecular weight 323.05 g/mol
  • CAS 100130-32-3
  • Appearance solid
  • Potency Ki = 31.6 nM; EC50 = 50 nM (histamine H4 receptor)
  • Available in small milligram pack sizes, including 10 mg
  • Intended for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.