Learn More
Medchemexpress LLC HY-101420 5mg Medchemexpress, VUF10460 CAS:1028327-66-3 Purity:>98%
SDP
Catalog No. 501932120
Encompass_Preferred
Medchemexpress, HY-101420 5mg VUF10460 CAS:1028327-66-3 VUF10460 is a non-imidazole histamine H4 receptor agonist; binds to rat H4 receptor with a pK i of 7.46. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2120 Supplier Medchemexpress LLC Supplier No. HY1014205MG
Add to Cart
Add to Cart
Medchemexpress, HY-101420 5mg VUF10460 CAS:1028327-66-3 VUF10460 is a non-imidazole histamine H4 receptor agonist; binds to rat H4 receptor with a pK i of 7.46. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.