Learn More
TARGETMOL CHEMICALS INC VUF11207 fumarate 10MG
SDP
Catalog No. 502876427
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. VUF11207 fumarate is an agonist of CXCR7 and a high-potency ligand of CXCR7 (pKi 8.1). Purity 99.5%
Catalog No. 50-287-6427 Supplier TARGETMOL CHEMICALS INC Supplier No. T1332410MG
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. VUF11207 fumarate is an agonist of CXCR7 and a high-potency ligand of CXCR7 (pKi 8.1). Purity 99.5%