Learn More
Medchemexpress LLC VUF11207 fumarate | 1785665-61-3 | 99.9% | 586.65 | 1 ML
SDP
Catalog No. 5000343968
Encompass_Preferred
VUF11207 fumarate is an agonist of CXCR7, a C-X-C motif chemokine receptor. It specifically binds to CXCR7 and has been shown to reduce CXCL12-mediated osteoclastogenesis and bone resorption. This reduction is achieved by inhibiting ERK phosphorylation. It is intended for research use only.
  • Binds specifically to CXCR7
  • Reduces osteoclastogenesis
  • Decreases bone resorption
  • Functions by inhibiting ERK phosphorylation
  • High purity: 99.9%
Catalog No. 50-003-43968 Supplier Medchemexpress LLC Supplier No. HY11031850MG
VUF11207 fumarate is an agonist of CXCR7, a C-X-C motif chemokine receptor. It specifically binds to CXCR7 and has been shown to reduce CXCL12-mediated osteoclastogenesis and bone resorption. This reduction is achieved by inhibiting ERK phosphorylation. It is intended for research use only.
  • Binds specifically to CXCR7
  • Reduces osteoclastogenesis
  • Decreases bone resorption
  • Functions by inhibiting ERK phosphorylation
  • High purity: 99.9%