Learn More
Medchemexpress LLC VUF11207 fumarate | 1785665-61-3 | 99.9% | 586.65 | 25 MG
SDP
Catalog No. 5000345917
Encompass_Preferred
VUF11207 fumarate is a CXCR7 agonist that specifically binds to CXCR7. It reduces CXCL12-mediated osteoclastogenesis and bone resorption by inhibiting ERK phosphorylation. This product is for research use only.
  • Specifically binds to CXCR7
  • Reduces CXCL12-mediated osteoclastogenesis
  • Inhibits bone resorption
  • Suppresses phosphorylation of ERK
  • Inhibits RANKL- and TNF-α-induced osteoclastogenesis
  • Inhibits LPS-induced osteoclastogenesis and bone resorption
Catalog No. 50-003-45917 Supplier Medchemexpress LLC Supplier No. 25MGHY110318
VUF11207 fumarate is a CXCR7 agonist that specifically binds to CXCR7. It reduces CXCL12-mediated osteoclastogenesis and bone resorption by inhibiting ERK phosphorylation. This product is for research use only.
  • Specifically binds to CXCR7
  • Reduces CXCL12-mediated osteoclastogenesis
  • Inhibits bone resorption
  • Suppresses phosphorylation of ERK
  • Inhibits RANKL- and TNF-α-induced osteoclastogenesis
  • Inhibits LPS-induced osteoclastogenesis and bone resorption