Learn More
Medchemexpress LLC VUF11207 fumarate | 1785665-61-3 | 99.9% | 586.65 | 50 MG
SDP
Catalog No. 5000344106
Encompass_Preferred
VUF11207 fumarate is a CXCR7 agonist that binds specifically to CXCR7. It reduces CXCL12-mediated osteoclastogenesis and bone resorption by inhibiting ERK phosphorylation.
  • Reduces CXCL12-mediated osteoclastogenesis and bone resorption by inhibiting ERK phosphorylation.
  • Binds specifically to CXCR7.
Catalog No. 50-003-44106 Supplier Medchemexpress LLC Supplier No. HY110318100MG
VUF11207 fumarate is a CXCR7 agonist that binds specifically to CXCR7. It reduces CXCL12-mediated osteoclastogenesis and bone resorption by inhibiting ERK phosphorylation.
  • Reduces CXCL12-mediated osteoclastogenesis and bone resorption by inhibiting ERK phosphorylation.
  • Binds specifically to CXCR7.