Learn More
Medchemexpress LLC VUF11207 fumarate | 1785665-61-3 | 99.9% | 586.65 g/mol | C31H39FN2O8 | 5 MG
SDP
Catalog No. 5000263465
Encompass_Preferred
VUF11207 fumarate is a research-grade small-molecule agonist of the chemokine receptor CXCR7 (ACKR3). It selectively binds CXCR7 and has been reported to reduce CXCL12-mediated osteoclastogenesis and bone resorption through inhibition of ERK phosphorylation. The compound is supplied for laboratory research use only.
  • Selective CXCR7 agonist with reported activity in CXCL12 signaling.
  • Reduces CXCL12-mediated osteoclastogenesis and bone resorption by inhibiting ERK phosphorylation.
  • High purity suitable for in vitro studies (99.94%).
  • Available as solid and as a 10 mM solution in DMSO; multiple solid package sizes available.
  • Intended for research use only; not for human or veterinary use.
  • Applicable to receptor pharmacology and cell-based signaling assays.
Catalog No. 50-002-63465 Supplier Medchemexpress LLC Supplier No. HY1103185MG
VUF11207 fumarate is a research-grade small-molecule agonist of the chemokine receptor CXCR7 (ACKR3). It selectively binds CXCR7 and has been reported to reduce CXCL12-mediated osteoclastogenesis and bone resorption through inhibition of ERK phosphorylation. The compound is supplied for laboratory research use only.
  • Selective CXCR7 agonist with reported activity in CXCL12 signaling.
  • Reduces CXCL12-mediated osteoclastogenesis and bone resorption by inhibiting ERK phosphorylation.
  • High purity suitable for in vitro studies (99.94%).
  • Available as solid and as a 10 mM solution in DMSO; multiple solid package sizes available.
  • Intended for research use only; not for human or veterinary use.
  • Applicable to receptor pharmacology and cell-based signaling assays.