Learn More
Medchemexpress LLC VULM 1457 100mg | 228544-65-8 | 100MG
SDP
Catalog No. 5000240017
Encompass_Preferred
VULM 1457 is a potent inhibitor of cholesterol acyltransferase (acyl-CoA) VULM1457 significantly reduces production and secretion of adrenomedullin (AM) and down-regulates AM receptors on human hepatoblastic cells VULM 1457 has remarkable hypolipidaemic activity and improves the overall myocardial ischaemia-reperfusion injury outcomes VULM 1457 has the potential for the research of diabetes mellitus and hypercholesterolaemia[1 [2
Catalog No. 50-002-40017 Supplier Medchemexpress LLC Supplier No. HY107571100MG
VULM 1457 is a potent inhibitor of cholesterol acyltransferase (acyl-CoA) VULM1457 significantly reduces production and secretion of adrenomedullin (AM) and down-regulates AM receptors on human hepatoblastic cells VULM 1457 has remarkable hypolipidaemic activity and improves the overall myocardial ischaemia-reperfusion injury outcomes VULM 1457 has the potential for the research of diabetes mellitus and hypercholesterolaemia[1 [2