Learn More
Medchemexpress LLC Vulm 1457 | 228544-65-8 | MFCD10687104 | >98.0% | 449.57 g/mol | C25H27N3O3S | 10 MG
SDP
Catalog No. 5000240114
Encompass_Preferred
VULM 1457 is a small-molecule research compound that potently inhibits cholesterol acyltransferase (ACAT). It reduces production and secretion of adrenomedullin and down-regulates adrenomedullin receptors in hepatoblastic cells, with reported hypolipidaemic activity and protective effects in myocardial ischaemia-reperfusion models. Provided as a high-purity research reagent (CAS 228544-65-8; MW 449.57 g/mol).
  • Potent ACAT inhibitor for metabolic enzyme studies.
  • Reduces adrenomedullin production and signaling.
  • Demonstrates hypolipidaemic activity in reported models.
  • Shows cardioprotective effects in ischaemia-reperfusion studies.
  • Available as a high-purity powder suitable for in vitro research.
Catalog No. 50-002-40114 Supplier Medchemexpress LLC Supplier No. HY10757110MG
VULM 1457 is a small-molecule research compound that potently inhibits cholesterol acyltransferase (ACAT). It reduces production and secretion of adrenomedullin and down-regulates adrenomedullin receptors in hepatoblastic cells, with reported hypolipidaemic activity and protective effects in myocardial ischaemia-reperfusion models. Provided as a high-purity research reagent (CAS 228544-65-8; MW 449.57 g/mol).
  • Potent ACAT inhibitor for metabolic enzyme studies.
  • Reduces adrenomedullin production and signaling.
  • Demonstrates hypolipidaemic activity in reported models.
  • Shows cardioprotective effects in ischaemia-reperfusion studies.
  • Available as a high-purity powder suitable for in vitro research.