Learn More
Cayman Chemical Vulpinic Acid, Purity: ≥98%, CAS: 521-52-8, 100 mg
SDP
Catalog No. 502986205
Encompass_Preferred
A lichen metabolite with diverse biological activities; active against various bacteria (MICs = 4-16 μg/ml); prevents UVB-induced apoptosis, cytotoxicity, and cytoskeletal modifications in HaCaT human keratinocytes from 25-800 μM; increases scratch wound healing of HaCaT cells; reduces hydrogen peroxide-induced production of ROS and cytotoxicity in HUVECs at 15 μM
Catalog No. 50-298-6205 Supplier Cayman Chemical Supplier No. 21685100 MG
A lichen metabolite with diverse biological activities; active against various bacteria (MICs = 4-16 μg/ml); prevents UVB-induced apoptosis, cytotoxicity, and cytoskeletal modifications in HaCaT human keratinocytes from 25-800 μM; increases scratch wound healing of HaCaT cells; reduces hydrogen peroxide-induced production of ROS and cytotoxicity in HUVECs at 15 μM