Learn More
Medchemexpress LLC Vupanorsen sodium | 2172852-12-7 | C296H414N86Na20O15P20S13 | 1 MG
SDP
Catalog No. 5000373842
Encompass_Preferred
Vupanorsen (sodium) is an N-acetyl galactosamine-conjugated antisense oligonucleotide that inhibits Angiopoietin-like 3 (ANGPTL3) protein synthesis. Vupanorsen (sodium) lowers triglycerides and atherogenic lipoproteins.
  • Inhibits Angiopoietin-like 3 (ANGPTL3) protein synthesis
  • Lowers triglycerides and atherogenic lipoproteins
Catalog No. 50-003-73842 Supplier Medchemexpress LLC Supplier No. HY153497A1MG
Vupanorsen (sodium) is an N-acetyl galactosamine-conjugated antisense oligonucleotide that inhibits Angiopoietin-like 3 (ANGPTL3) protein synthesis. Vupanorsen (sodium) lowers triglycerides and atherogenic lipoproteins.
  • Inhibits Angiopoietin-like 3 (ANGPTL3) protein synthesis
  • Lowers triglycerides and atherogenic lipoproteins