Learn More
Medchemexpress LLC HY-122610 5mg Medchemexpress, VUT-MK142 CAS:1313491-22-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1226105MG
Encompass_Preferred
Medchemexpress, HY-122610 5mg VUT-MK142 CAS:1313491-22-3 VUT-MK142 is a potent new cardiomyogenic synthetic agent promoting the differentiation of pre-cardiac mesoderm into cardiomyocytes, which may be useful to differentiate stem cells into cardiomyocytes for cardiac repair. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0991