Learn More
Vector Laboratories Vicia Villosa Lectin (VVL, VVA), Unconjugated, 5mg
SDP
Supplier:  Vector Laboratories L12305
Encompass
Recommended Usage: Although many buffers can be employed for reconstituting this lectin, we recommend 10 mM HEPES buffered saline, pH 8.5, 0.1 mM CaCl2.For preserving solutions stored at 4 ºC, 0.08% sodium azide can be used. Recommended Storage: 2-8 °C 5 mg , Glycobiology.
Catalog No. 50-204-5674