Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 7450MSM5P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: VWF/1465. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: HUVEC cells or Tonsil. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant fragment of human vWF protein (aa1815-1939). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 250kDa.Chromosome: 12p13.31. Gene Symbol: VWF. Gene ID: 7450. Ensemble: ENSG00000110799. Uniprot: NP_000543.
Catalog No. 50-188-5282