Learn More
Genetex Inc VWF Monoclonal Antibody [MSVA-521R] HistoMAX™ (GTX04412) GeneTex - IHC-Optimized Antibody.
SDP
Catalog No. NC3666785
Encompass_Preferred
The VWF antibody [MSVA-521R] HistoMAX™ (GTX04412) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human skeletal muscle tissue using GTX04412 VWF antibody [MSVA-521R] HistoMAX. In skeletal muscle endothelial VWF immunostaining is strong in postcapillary venules and somewhat weaker in capillaries.
Catalog No. NC3666785 Supplier Genetex Inc Supplier No. GTX04412
The VWF antibody [MSVA-521R] HistoMAX™ (GTX04412) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human skeletal muscle tissue using GTX04412 VWF antibody [MSVA-521R] HistoMAX. In skeletal muscle endothelial VWF immunostaining is strong in postcapillary venules and somewhat weaker in capillaries.