Learn More
Cell Signaling Technology VWF Monoclonal Antibody, Rabbit, Human, 20 μl
SDP
Catalog No. 502732262
Encompass
VWF (D8L8G) Rabbit Monoclonal Antibody recognizes endogenous levels of total VWF protein, detecting both the VWF precursor and mature VWF. The antibody is produced by immunizing animals with recombinant human VWF protein.
  • Recognizes endogenous total VWF protein
  • Detects both precursor and mature VWF
  • Validated for western blotting, immunoprecipitation, and immunohistochemistry (paraffin) applications
  • Host species is rabbit
  • Reacts with human samples
Catalog No. 50-273-2262 Supplier Cell Signaling Technology Supplier No. 65707T
VWF (D8L8G) Rabbit Monoclonal Antibody recognizes endogenous levels of total VWF protein, detecting both the VWF precursor and mature VWF. The antibody is produced by immunizing animals with recombinant human VWF protein.
  • Recognizes endogenous total VWF protein
  • Detects both precursor and mature VWF
  • Validated for western blotting, immunoprecipitation, and immunohistochemistry (paraffin) applications
  • Host species is rabbit
  • Reacts with human samples