Learn More
Abcam Human Von Willebrand Factor ELISA Kit (VWF), 1 x 96 Tests
Supplier:  Abcam AB1089181X96TESTS
Encompass_Preferred
Human Von Willebrand Factor ELISA Kit (VWF) is a Sandwich (quantitative) ELISA kit for the measurement of Human Von Willebrand Factor (VWF) in Human in Plasma, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0575231