Learn More
Andwin Scientific Disposable Pipetting Reservoirs, 100 mL, Sterile, Individually Wrapped, 100/cs
SDP
Catalog No. NC0637709
Encompass_Preferred
Disposable Pipetting Reservoirs are designed for use in multichannel pipetting applications, and will accommodate all types of pipettors up to 12-channel width. Pipetting reservoirs feature a trough design, graduation marks on interior walls, and pour-off spouts on each corner. All reservoirs are lot-certified, nonpyrogenic, and DNase- and RNase-free.
Catalog No. NC0637709 Supplier Andwin Scientific Supplier No. 89094658
Disposable Pipetting Reservoirs are designed for use in multichannel pipetting applications, and will accommodate all types of pipettors up to 12-channel width. Pipetting reservoirs feature a trough design, graduation marks on interior walls, and pour-off spouts on each corner. All reservoirs are lot-certified, nonpyrogenic, and DNase- and RNase-free.